Assay Details
| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204962.1 | NP_990293.1 | 1 - 2 | 179 | 77 | |
| GenBank mRNA | U44473.1 | - | 1 - 2 | 179 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CAMLG| Gene Symbol: CAMLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204962.1 | NP_990293.1 | 1 - 2 | 179 | 77 | |
| GenBank mRNA | U44473.1 | - | 1 - 2 | 179 | 77 | |
Set Membership: |
|