Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204967.1 | NP_990298.1 | 1196 | 71 | ||
| GenBank mRNA | U34977.1 | - | 1196 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FMOD| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204967.1 | NP_990298.1 | 1196 | 71 | ||
| GenBank mRNA | U34977.1 | - | 1196 | 71 | ||
Set Membership: |
|