Assay Details
| Gene Symbol: KIF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204975.1 | NP_990306.1 | 24 - 25 | 3056 | 75 | |
| XM_015278192.2 | XP_015133678.1 | 24 - 25 | 3113 | 75 | ||
| GenBank mRNA | U04821.1 | - | 7 - 8 | 808 | 75 | |
| U18309.1 | - | 24 - 25 | 3056 | 75 | ||
More Information
Set Membership: |
|

KIF4B
Genomic Map