Assay Details
| Gene Symbol: VDHAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204976.2 | NP_990307.2 | 9 - 10 | 1312 | 119 | |
| GenBank mRNA | U00694.1 | - | 9 - 10 | 1345 | 119 | |
More Information
Set Membership: |
|

VDHAP
Genomic Map