Assay Details
| Gene Symbol: GFRA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204991.1 | NP_990322.1 | 1 - 2 | 39 | 70 | |
| XM_015286039.2 | XP_015141525.1 | 1 - 2 | 520 | 70 | ||
| GenBank mRNA | AF045162.1 | - | 1 - 2 | 39 | 70 | |
More Information
Set Membership: |
|

GFRA4
Genomic Map