Assay Details
| Gene Symbol: DEFB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201399.1 | NP_001188328.1 | 3 - 4 | 335 | 103 | |
| NM_204992.2 | NP_990323.2 | 3 - 4 | 271 | 103 | ||
| XM_015285091.2 | XP_015140577.1 | 3 - 4 | 288 | 103 | ||
| GenBank mRNA | AF033336.1 | - | 2 - 3 | 211 | 102 | |
| BQ484799.1 | - | 3 - 4 | 251 | 104 | ||
| CF250963.1 | - | 3 - 4 | 337 | 103 | ||
More Information
Set Membership: |
|

DEFB4A
Genomic Map