Assay Details
| Gene Symbol: GJA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204997.1 | NP_990328.1 | 1 | 850 | 110 | |
| XM_015295800.2 | XP_015151286.1 | 5 | 1347 | 110 | ||
| XM_015295808.2 | XP_015151294.1 | 3 | 1070 | 110 | ||
| XM_015295814.2 | XP_015151300.1 | 4 | 1205 | 110 | ||
| XM_015295822.2 | XP_015151308.1 | 3 | 6126 | 110 | ||
| XM_015295825.2 | XP_015151311.1 | 2 | 993 | 110 | ||
More Information
Set Membership: |
|

GJA8
Genomic Map