Assay Details
| Gene Symbol: RAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205016.1 | NP_990347.1 | 1 - 2 | 60 | 65 | |
| XM_025141545.1 | XP_024997313.1 | 1 - 2 | 149 | 65 | ||
| GenBank mRNA | U79756.1 | - | 1 - 2 | 60 | 65 | |
More Information
Set Membership: |
|

RAC3
Genomic Map