Assay Details
| Gene Symbol: CDC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205048.1 | NP_990379.1 | 2 - 3 | 338 | 119 | |
| XM_015296825.2 | XP_015152311.1 | 2 - 3 | 189 | 119 | ||
| XM_015296826.2 | XP_015152312.1 | 2 - 3 | 239 | 119 | ||
| XM_015296827.2 | XP_015152313.1 | 2 - 3 | 206 | 119 | ||
| XM_025142484.1 | XP_024998252.1 | 2 - 3 | 315 | 119 | ||
| GenBank mRNA | U40848.1 | - | 2 - 3 | 338 | 119 | |
More Information
Set Membership: |
|

CDC42
Genomic Map