Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205062.1 | NP_990393.1 | 5 - 6 | 716 | 72 | |
| GenBank mRNA | AJ001912.1 | - | 5 - 6 | 716 | 72 | |
More Information
Set Membership: |
|

LRPAP1
Genomic Map