Assay Details
| Gene Symbol: LDB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205070.1 | NP_990401.1 | 6 - 7 | 580 | 106 | |
| XM_015288581.2 | XP_015144067.2 | 6 - 7 | 721 | 106 | ||
| XM_015288582.2 | XP_015144068.1 | 6 - 7 | 632 | 106 | ||
| XM_015288583.2 | XP_015144069.1 | 6 - 7 | 514 | 106 | ||
| GenBank mRNA | AB250385.1 | - | 5 - 6 | 419 | 106 | |
| AF026037.1 | - | 6 - 7 | 580 | 106 | ||
| AF026038.1 | - | 6 - 7 | 564 | 106 | ||
| AF069991.1 | - | 5 - 6 | 412 | 106 | ||
More Information
Set Membership: |
|

LDB1
Genomic Map