Assay Details
| Gene Symbol: ENO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205120.1 | NP_990451.1 | 11 - 12 | 1263 | 57 | |
| XM_015296833.2 | XP_015152319.2 | 11 - 12 | 1328 | 57 | ||
| XM_015296834.2 | XP_015152320.2 | 11 - 12 | 1259 | 57 | ||
| XM_025142487.1 | XP_024998255.1 | 11 - 12 | 1266 | 57 | ||
| GenBank mRNA | D37900.1 | - | 11 - 12 | 1263 | 57 | |
More Information
Set Membership: |
|

ENO1
Genomic Map