Assay Details
| Gene Symbol: IFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205149.1 | NP_990480.1 | 3 - 4 | 476 | 74 | |
| GenBank mRNA | AF424744.1 | - | 3 - 4 | 371 | 74 | |
| AJ634956.1 | - | 3 - 4 | 370 | 74 | ||
| AY501004.1 | - | 3 - 4 | 377 | 74 | ||
| DQ470471.1 | - | 3 - 4 | 371 | 74 | ||
| EU931580.1 | - | 1 - 2 | 101 | 74 | ||
| FJ788637.1 | - | 3 - 4 | 371 | 74 | ||
| GQ246226.1 | - | 3 - 4 | 371 | 74 | ||
| GQ421600.1 | - | 3 - 4 | 371 | 74 | ||
| HQ739082.1 | - | 2 - 3 | 228 | 74 | ||
| JN942588.1 | - | 2 - 3 | 228 | 74 | ||
| U27465.1 | - | 3 - 4 | 467 | 74 | ||
| X99774.1 | - | 476 | 74 | |||
More Information
Set Membership: |
|

IFNG
Genomic Map