Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205175.3 | NP_990506.2 | 2 - 3 | 152 | 62 | |
| XM_015275225.2 | XP_015130711.1 | 2 - 3 | 62 | |||
| GenBank mRNA | AY539829.1 | - | 2 - 3 | 152 | 62 | |
| AY539830.1 | - | 2 - 3 | 152 | 62 | ||
More Information
Set Membership: |
|

CRYBB2
Genomic Map