Assay Details
| Gene Symbol: LMNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205286.1 | NP_990617.1 | 8 - 9 | 1580 | 62 | |
| GenBank mRNA | X16878.1 | - | 1580 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LMNB1| Gene Symbol: LMNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205286.1 | NP_990617.1 | 8 - 9 | 1580 | 62 | |
| GenBank mRNA | X16878.1 | - | 1580 | 62 | ||
Set Membership: |
|