Assay Details
| Gene Symbol: HRAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205292.1 | NP_990623.1 | 3 - 4 | 542 | 72 | |
| GenBank mRNA | HQ439436.1 | - | 3 - 4 | 536 | 72 | |
| X03578.1 | - | 3 - 4 | 577 | 72 | ||
More Information
Set Membership: |
|

HRAS
Genomic Map