Assay Details
| Gene Symbol: H3F3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205296.1 | NP_990627.1 | 3 - 4 | 393 | 92 | |
| GenBank mRNA | Y00392.1 | - | 393 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H3F3B| Gene Symbol: H3F3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205296.1 | NP_990627.1 | 3 - 4 | 393 | 92 | |
| GenBank mRNA | Y00392.1 | - | 393 | 92 | ||
Set Membership: |
|