Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205297.1 | NP_990628.1 | 4 - 5 | 479 | 129 | |
| GenBank mRNA | X60795.1 | - | 479 | 129 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPCS3| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205297.1 | NP_990628.1 | 4 - 5 | 479 | 129 | |
| GenBank mRNA | X60795.1 | - | 479 | 129 | ||
Set Membership: |
|