Assay Details
| Gene Symbol: CKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205310.1 | NP_990641.1 | 3 - 4 | 528 | 77 | |
| XM_015287304.2 | XP_015142790.1 | 4 - 5 | 555 | 77 | ||
| XM_025150611.1 | XP_025006379.1 | 5 - 6 | 766 | 77 | ||
| GenBank mRNA | BX932948.1 | - | 4 - 5 | 539 | 77 | |
| X03509.1 | - | 528 | 77 | |||
More Information
Set Membership: |
|

CKB
Genomic Map