Assay Details
| Gene Symbol: RPLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205322.1 | NP_990653.1 | 3 - 4 | 341 | 159 | |
| GenBank mRNA | X13876.1 | - | 341 | 159 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPLP1| Gene Symbol: RPLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205322.1 | NP_990653.1 | 3 - 4 | 341 | 159 | |
| GenBank mRNA | X13876.1 | - | 341 | 159 | ||
Set Membership: |
|