Assay Details
| Gene Symbol: NPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205473.1 | NP_990804.1 | 3 - 4 | 314 | 72 | |
| GenBank mRNA | CR390268.1 | - | 3 - 4 | 514 | 72 | |
| M87294.1 | - | 3 - 4 | 314 | 72 | ||
More Information
Set Membership: |
|

NPY
Genomic Map