Assay Details
| Gene Symbol: IL8L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205498.1 | NP_990829.1 | 3 - 4 | 363 | 69 | |
| GenBank mRNA | HM179639.1 | - | 3 - 4 | 306 | 69 | |
| J02975.1 | - | 3 - 4 | 340 | 69 | ||
| M16199.1 | - | 3 - 4 | 363 | 69 | ||
More Information
Set Membership: |
|

IL8L2
Genomic Map