Assay Details
| Gene Symbol: PDCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030812.1 | NP_001025983.1 | 2 - 3 | 217 | 70 | |
| GenBank mRNA | AJ719844.1 | - | 2 - 3 | 217 | 70 | |
More Information
Set Membership: |
|

PDCL3
Genomic Map