Assay Details
| Gene Symbol: ACOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030821.1 | NP_001025992.1 | 3 - 4 | 321 | 65 | |
| XM_015275372.2 | XP_015130858.1 | 4 - 5 | 439 | 65 | ||
| GenBank mRNA | AJ720739.1 | - | 3 - 4 | 321 | 65 | |
More Information
Set Membership: |
|

ACOD1
Genomic Map