Assay Details
| Gene Symbol: SPPL2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030933.1 | NP_001026104.1 | 2 - 3 | 281 | 86 | |
| XM_015299708.2 | XP_015155194.1 | 2 - 3 | 86 | |||
| GenBank mRNA | AJ851767.1 | - | 2 - 3 | 281 | 86 | |
More Information
Set Membership: |
|

SPPL2C
Genomic Map