Assay Details
| Gene Symbol: STK17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030995.1 | NP_001026166.1 | 5 - 6 | 806 | 86 | |
| XM_015281562.2 | XP_015137048.1 | 5 - 6 | 616 | 86 | ||
| GenBank mRNA | AJ719290.1 | - | 5 - 6 | 835 | 86 | |
More Information
Set Membership: |
|

STK17A
Genomic Map