Assay Details
| Gene Symbol: MAPRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031017.1 | NP_001026188.1 | 6 - 7 | 980 | 72 | |
| XM_015282587.2 | XP_015138073.2 | 7 - 8 | 1009 | 72 | ||
| XM_015282589.2 | XP_015138075.2 | 7 - 8 | 962 | 72 | ||
| XM_015282590.2 | XP_015138076.2 | 7 - 8 | 899 | 72 | ||
| XM_025147305.1 | XP_025003073.1 | 6 - 7 | 989 | 72 | ||
| GenBank mRNA | AJ720083.1 | - | 6 - 7 | 980 | 72 | |
More Information
Set Membership: |
|

MAPRE2
Genomic Map