Assay Details
| Gene Symbol: UBXN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031023.1 | NP_001026194.1 | 7 - 8 | 806 | 67 | |
| GenBank mRNA | AJ719732.1 | - | 7 - 8 | 806 | 67 | |
More Information
Set Membership: |
|

UBXN2B
Genomic Map