Assay Details
| Gene Symbol: SDCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031024.1 | NP_001026195.1 | 2 - 3 | 185 | 69 | |
| XM_015282672.2 | XP_015138158.1 | 2 - 3 | 216 | 69 | ||
| XM_025147307.1 | XP_025003075.1 | 2 - 3 | 95 | 69 | ||
| XM_025147308.1 | XP_025003076.1 | 2 - 3 | 135 | 69 | ||
| XM_025147309.1 | XP_025003077.1 | 2 - 3 | 145 | 69 | ||
| GenBank mRNA | AJ721106.1 | - | 2 - 3 | 185 | 69 | |
More Information
Set Membership: |
|

SDCBP
Genomic Map