Assay Details
| Gene Symbol: KLHL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031116.1 | NP_001026287.1 | 6 - 7 | 672 | 120 | |
| XM_015278239.2 | XP_015133725.1 | 5 - 6 | 625 | 120 | ||
| XM_015278240.2 | XP_015133726.1 | 4 - 5 | 723 | 120 | ||
| XM_015278241.2 | XP_015133727.1 | 4 - 5 | 865 | 120 | ||
| XM_015278242.2 | XP_015133728.1 | 5 - 6 | 622 | 120 | ||
| XM_025149555.1 | XP_025005323.1 | 6 - 7 | 652 | 120 | ||
| GenBank mRNA | AJ719801.1 | - | 6 - 7 | 672 | 120 | |
More Information
Set Membership: |
|

KLHL9
Genomic Map