Assay Details
| Gene Symbol: NELFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031167.1 | NP_001026338.1 | 9 - 10 | 1737 | 63 | |
| XM_025149713.1 | XP_025005481.1 | 10 - 11 | 1567 | 63 | ||
| XM_025149714.1 | XP_025005482.1 | 9 - 10 | 1477 | 63 | ||
| XM_025149715.1 | XP_025005483.1 | 8 - 9 | 1315 | 63 | ||
| GenBank mRNA | AJ851513.1 | - | 9 - 10 | 1737 | 63 | |
More Information
Set Membership: |
|

NELFA
Genomic Map