Assay Details
| Gene Symbol: VPS51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031172.1 | NP_001026343.1 | 9 - 10 | 2136 | 73 | |
| XM_015286198.2 | XP_015141684.1 | 8 - 9 | 2021 | 73 | ||
| GenBank mRNA | AJ720609.1 | - | 9 - 10 | 2136 | 73 | |
| CR390974.1 | - | 2 - 3 | 123 | 73 | ||
More Information
Set Membership: |
|

VPS51
Genomic Map