Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031227.1 | NP_001026398.1 | 3 - 4 | 724 | 61 | |
| XM_015288218.2 | XP_015143704.1 | 4 - 5 | 624 | 61 | ||
| GenBank mRNA | AJ719337.1 | - | 3 - 4 | 724 | 61 | |
More Information
Set Membership: |
|

AP3M1
Genomic Map