Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031233.1 | NP_001026404.1 | 2 - 3 | 701 | 65 | |
| GenBank mRNA | AJ719866.1 | - | 2 - 3 | 701 | 65 | |
More Information
Set Membership: |
|

TRIM8
Genomic Map