Assay Details
| Gene Symbol: TAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031235.1 | NP_001026406.1 | 7 - 8 | 1634 | 79 | |
| GenBank mRNA | AJ851663.1 | - | 7 - 8 | 1634 | 79 | |
More Information
Set Membership: |
|

TAF5
Genomic Map