Assay Details
| Gene Symbol: UMPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031260.1 | NP_001026431.1 | 1 - 2 | 175 | 69 | |
| GenBank mRNA | AJ719322.1 | - | 1 - 2 | 175 | 69 | |
More Information
Set Membership: |
|

UMPS
Genomic Map