Assay Details
| Gene Symbol: AMACR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031448.1 | NP_001026619.1 | 4 - 5 | 784 | 97 | |
| GenBank mRNA | AJ719871.1 | - | 4 - 5 | 784 | 97 | |
More Information
Set Membership: |
|

AMACR
Genomic Map