Assay Details
| Gene Symbol: SUDS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031471.1 | NP_001026642.1 | 9 - 10 | 675 | 81 | |
| XM_015275338.2 | XP_015130824.1 | 9 - 10 | 3776 | 81 | ||
| GenBank mRNA | AJ720986.1 | - | 9 - 10 | 675 | 81 | |
More Information
Set Membership: |
|

SUDS3
Genomic Map