Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031475.1 | NP_001026646.1 | 2 - 3 | 299 | 62 | |
| XM_015279526.2 | XP_015135012.1 | 2 - 3 | 326 | 62 | ||
| GenBank mRNA | AJ719979.1 | - | 2 - 3 | 299 | 62 | |
More Information
Set Membership: |
|

ARRDC1
Genomic Map