Assay Details
| Gene Symbol: RAB8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031504.1 | NP_001026675.1 | 12 - 13 | 1738 | 87 | |
| GenBank mRNA | AJ851430.1 | - | 12 - 13 | 1738 | 87 | |
| BX930100.2 | - | 5 - 6 | 422 | 87 | ||
More Information
Set Membership: |
|

RAB8A
Genomic Map