Assay Details
| Gene Symbol: PIGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031522.2 | NP_001026693.2 | 1 | 1057 | 81 | |
| GenBank mRNA | AJ851770.1 | - | 1 | 1057 | 81 | |
| BU222932.1 | - | 1 | 70 | 81 | ||
| BX271414.4 | - | 1 | 452 | 81 | ||
More Information
Set Membership: |
|

PIGM
Genomic Map