Assay Details
| Gene Symbol: ACP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031548.2 | NP_001026719.2 | 8 - 9 | 914 | 66 | |
| GenBank mRNA | AJ720873.1 | - | 8 - 9 | 914 | 66 | |
| BG713467.1 | - | 5 - 6 | 557 | 66 | ||
More Information
Set Membership: |
|

ACP2
Genomic Map