Assay Details
| Gene Symbol: ARPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031561.1 | NP_001026732.1 | 3 - 4 | 436 | 130 | |
| GenBank mRNA | AJ719279.1 | - | 3 - 4 | 436 | 130 | |
| CR385694.1 | - | 3 - 4 | 432 | 130 | ||
| CR389543.1 | - | 3 - 4 | 432 | 130 | ||
More Information
Set Membership: |
|

ARPC5
Genomic Map