Assay Details
| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031568.2 | NP_001026739.2 | 6 - 7 | 601 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNG| Gene Symbol: CHRNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031568.2 | NP_001026739.2 | 6 - 7 | 601 | 71 | |
Set Membership: |
|