Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030663.1 | NP_001025834.1 | 4 - 5 | 609 | 61 | |
| XM_015275297.2 | XP_015130783.1 | 4 - 5 | 612 | 61 | ||
| XM_015275298.2 | XP_015130784.1 | 4 - 5 | 689 | 61 | ||
| XM_025155462.1 | XP_025011230.1 | 5 - 6 | 702 | 61 | ||
| XM_025155463.1 | XP_025011231.1 | 4 - 5 | 586 | 61 | ||
| XM_025155464.1 | XP_025011232.1 | 4 - 5 | 599 | 61 | ||
| GenBank mRNA | AJ851380.1 | - | 4 - 5 | 609 | 61 | |
More Information
Set Membership: |
|

HVCN1
Genomic Map