Assay Details
| Gene Symbol: BG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030670.1 | NP_001025841.1 | 33 - 34 | 1302 | 124 | |
| GenBank mRNA | M61864.1 | - | 33 - 34 | 1302 | 124 | |
| MH156617.1 | - | 33 - 34 | 1313 | 124 | ||
| MH156619.1 | - | 33 - 34 | 1313 | 124 | ||
| MH156621.1 | - | 30 - 31 | 1464 | 124 | ||
| Gene Symbol: LOC425214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155970.1 | XP_025011738.1 | 26 - 27 | 690 | 103 | |
More Information
Set Membership: |
|

BG8
Genomic Map