Assay Details
| Gene Symbol: BG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030670.1 | NP_001025841.1 | 12 - 13 | 863 | 149 | |
| GenBank mRNA | M61864.1 | - | 12 - 13 | 863 | 149 | |
| MH156615.1 | - | 11 - 12 | 874 | 149 | ||
| MH156616.1 | - | 11 - 12 | 874 | 149 | ||
| MH156617.1 | - | 12 - 13 | 874 | 149 | ||
| MH156618.1 | - | 12 - 13 | 874 | 259 | ||
| MH156619.1 | - | 12 - 13 | 874 | 149 | ||
| MH156620.1 | - | 12 - 13 | 874 | 149 | ||
| MH156623.1 | - | 12 - 13 | 874 | 149 | ||
| MH156641.1 | - | 11 - 12 | 874 | 149 | ||
More Information
Set Membership: |
|

BG8
Genomic Map