Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030701.1 | NP_001025872.1 | 10 - 11 | 1724 | 89 | |
| XM_015295143.2 | XP_015150629.1 | 10 - 11 | 1828 | 89 | ||
| XM_015295144.2 | XP_015150630.1 | 11 - 12 | 1897 | 89 | ||
| XM_015295145.2 | XP_015150631.1 | 10 - 11 | 2286 | 89 | ||
| XM_015295146.2 | XP_015150632.1 | 10 - 11 | 1756 | 89 | ||
| XM_015295147.2 | XP_015150633.1 | 9 - 10 | 955 | 89 | ||
| XM_015295148.2 | XP_015150634.1 | 9 - 10 | 1029 | 89 | ||
| XM_015295149.2 | XP_015150635.1 | 9 - 10 | 1054 | 89 | ||
| XM_025141592.1 | XP_024997360.1 | 11 - 12 | 2018 | 89 | ||
| GenBank mRNA | AJ851568.1 | - | 10 - 11 | 1724 | 89 | |
More Information
Set Membership: |
|

SEPTIN9
Genomic Map