Assay Details
| Gene Symbol: SEPTIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030701.1 | NP_001025872.1 | 8 - 9 | 1574 | 59 | |
| XM_015295143.2 | XP_015150629.1 | 8 - 9 | 1678 | 59 | ||
| XM_015295144.2 | XP_015150630.1 | 9 - 10 | 1747 | 59 | ||
| XM_015295145.2 | XP_015150631.1 | 8 - 9 | 2136 | 59 | ||
| XM_015295146.2 | XP_015150632.1 | 8 - 9 | 1606 | 59 | ||
| XM_015295147.2 | XP_015150633.1 | 7 - 8 | 805 | 59 | ||
| XM_015295148.2 | XP_015150634.1 | 7 - 8 | 879 | 59 | ||
| XM_015295149.2 | XP_015150635.1 | 7 - 8 | 904 | 59 | ||
| XM_025141592.1 | XP_024997360.1 | 9 - 10 | 1868 | 59 | ||
| GenBank mRNA | AJ851568.1 | - | 8 - 9 | 1574 | 59 | |
More Information
Set Membership: |
|

SEPTIN9
Genomic Map