Assay Details
| Gene Symbol: JMJD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030703.1 | NP_001025874.1 | 6 - 7 | 1304 | 59 | |
| GenBank mRNA | AJ719379.1 | - | 6 - 7 | 1304 | 59 | |
More Information
Set Membership: |
|

JMJD6
Genomic Map